missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Piperidyl)-1H-1,2,3-benzotriazole hydrochloride, ≥90%, Thermo Scientific™

Catalog Number p-2694403
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
79098-80-9
C11H15ClN4
238.719
MFCD00831110
SIZIQJKLJZWLLD-UHFFFAOYSA-N
1-4-piperidyl-1h-1,2,3-benzotriazole hydrochloride, 1-piperidin-4-ylbenzotriazole hydrochloride, 1-piperidin-4-yl benzotriazole hydrochloride, 1-piperidin-4-yl-1h-benzotriazole hydrochloride, 1-piperidin-4-yl-1,2,3-benzotriazole hydrochloride, c11h14n4.hcl, 4-piperidylbenzotriazole, chloride, 4-benzotriazol-1-yl-piperidine hcl salt, 4-benzotriazol-1-ylpiperidine hydrochloride, 1-4-piperidinyl benzotriazole hydrochloride
2779380
1-piperidin-4-ylbenzotriazole;hydrochloride
C1CNCCC1N2C3=CC=CC=C3N=N2.Cl
This item is not returnable. View return policy
79098-80-9
C11H15ClN4
238.719
MFCD00831110
SIZIQJKLJZWLLD-UHFFFAOYSA-N
1-4-piperidyl-1h-1,2,3-benzotriazole hydrochloride, 1-piperidin-4-ylbenzotriazole hydrochloride, 1-piperidin-4-yl benzotriazole hydrochloride, 1-piperidin-4-yl-1h-benzotriazole hydrochloride, 1-piperidin-4-yl-1,2,3-benzotriazole hydrochloride, c11h14n4.hcl, 4-piperidylbenzotriazole, chloride, 4-benzotriazol-1-yl-piperidine hcl salt, 4-benzotriazol-1-ylpiperidine hydrochloride, 1-4-piperidinyl benzotriazole hydrochloride
2779380
1-piperidin-4-ylbenzotriazole;hydrochloride
C1CNCCC1N2C3=CC=CC=C3N=N2.Cl
CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 238.72
Percent Purity 90%
Chemical Name or Material 1-(4-Piperidyl)-1H-1, 2, 3-benzotriazole hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'