missing translation for 'onlineSavingsMsg'
Learn More

1,4-Phenylene diisothiocyanate, 98%, Thermo Scientific Chemicals

Catalog Number p-7045193
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4044-65-9
C8H4N2S2
192.254
MFCD00004811
OMWQUXGVXQELIX-UHFFFAOYSA-N
bitoscanate, 1,4-phenylene diisothiocyanate, jonit, p-phenylene diisothiocyanate, benzene, 1,4-diisothiocyanato, bitoscanat, bitoscanate inn, phenylene thiocyanate, bitoscanatum latin, 1,4-phenylenediisothiocyanate
19958
1,4-diisothiocyanatobenzene
C1=CC(=CC=C1N=C=S)N=C=S
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4044-65-9
C8H4N2S2
192.254
MFCD00004811
OMWQUXGVXQELIX-UHFFFAOYSA-N
bitoscanate, 1,4-phenylene diisothiocyanate, jonit, p-phenylene diisothiocyanate, benzene, 1,4-diisothiocyanato, bitoscanat, bitoscanate inn, phenylene thiocyanate, bitoscanatum latin, 1,4-phenylenediisothiocyanate
19958
1,4-diisothiocyanatobenzene
C1=CC(=CC=C1N=C=S)N=C=S

It is used in the treatment of hookworms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the treatment of hookworms.

Solubility
Hydrolyzes in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 128°C to 133°C
Odor Irritating
Quantity 1 g
UN Number UN2923
Beilstein 2414340
Sensitivity Moisture sensitive
Merck Index 14,1308
Solubility Information Hydrolyzes in water.
Formula Weight 192.26
Percent Purity 98%
Chemical Name or Material 1,4-Phenylene diisothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'