missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,4-Oxathiane 4,4-dioxide, 99%

Brand:  Alfa Aesar™ A10210.06

Code : NEW

Additional Details : CAS Number : 107-61-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11489532

  • £14.40 / 5g


Estimated Shipment date
from Supplier 22-06-2018
Add to basket

Specifications

Chemical Identifiers

1,4-Oxathiane 4,4-dioxide
C4H8O3S
MFCD00011683
4,4-Dioxo-1,4-oxathiane; 1,4-Thioxane 1,1-dioxide
107-61-9
136.16
110609

Specifications

1,4-Oxathiane 4,4-dioxide
99%
136.16
110609
131° to 133°C
5g
107-61-9
C4H8O3S
MFCD00011683
4,4-Dioxo-1,4-oxathiane; 1,4-Thioxane 1,1-dioxide
Odorless
Videos
SDS