missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,4-Oxathiane 4,4-dioxide, 99%

Brand:  Alfa Aesar™ A10210.22

Code : NEW

Additional Details : CAS Number : 107-61-9 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11409542

  • £154.00 / 100g


Estimated Shipment date
from Supplier 07-02-2018
Add to basket

Specifications

Chemical Identifiers

1,4-Oxathiane 4,4-dioxide
C4H8O3S
MFCD00011683
4,4-Dioxo-1,4-oxathiane; 1,4-Thioxane 1,1-dioxide
107-61-9
136.16
110609

Specifications

1,4-Oxathiane 4,4-dioxide
99%
136.16
110609
131° to 133°C
100g
107-61-9
C4H8O3S
MFCD00011683
4,4-Dioxo-1,4-oxathiane; 1,4-Thioxane 1,1-dioxide
Odorless
Videos
SDS