missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Nitrophenyl)pyrrolidine, 97%

Brand:  Alfa Aesar™ H63958.06

Code : NEW

Additional Details : CAS Number : 10220-22-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15400959

  • £149.46 / 5g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Nitrophenyl)pyrrolidine
10220-22-1
192.21
MFCD00020819

Specifications

1-(4-Nitrophenyl)pyrrolidine
Solid
192.21
5g
10220-22-1
97%
MFCD00020819
Videos
SDS