missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Nitrophenyl)homopiperazine, 97%

Brand:  Alfa Aesar™ H63914.03

Code : NEW

Additional Details : CAS Number : 214124-83-1 Weight : 0.00100kg Transport : UN number : 3259 Chem class : 8 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15400879

  • £59.78 / 1g


Estimated Shipment date
from Supplier 25-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Nitrophenyl)homopiperazine
221.26
1-(4-Nitrophenyl)-1,4-diazepane
214124-83-1
UN3259

Specifications

1-(4-Nitrophenyl)homopiperazine
Solid
221.26
1g
214124-83-1
97%
UN3259
1-(4-Nitrophenyl)-1,4-diazepane
Videos
SDS