missing translation for 'onlineSavingsMsg'
Learn More

1,4-Naphthoquinone, 99%, contains up to 6% water, Thermo Scientific Chemicals

Catalog Number p-170789
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
130-15-4, 7732-18-5
C10H6O2
158.16
MFCD00001676
FRASJONUBLZVQX-UHFFFAOYSA-N
1,4-naphthoquinone, 1,4-naphthalenedione, p-naphthoquinone, naphthoquinone, alpha-naphthoquinone, 1,4-naphthylquinone, usaf cy-10, 1,4-dihydronaphthalene-1,4-dione, 1,4-dihydro-1,4-diketonaphthalene, 1,4-naftochinon
8530
CHEBI:27418
naphthalene-1,4-dione
C1=CC=C2C(=O)C=CC(=O)C2=C1
This item is not returnable. View return policy
130-15-4, 7732-18-5
C10H6O2
158.16
MFCD00001676
FRASJONUBLZVQX-UHFFFAOYSA-N
1,4-naphthoquinone, 1,4-naphthalenedione, p-naphthoquinone, naphthoquinone, alpha-naphthoquinone, 1,4-naphthylquinone, usaf cy-10, 1,4-dihydronaphthalene-1,4-dione, 1,4-dihydro-1,4-diketonaphthalene, 1,4-naftochinon
8530
CHEBI:27418
naphthalene-1,4-dione
C1=CC=C2C(=O)C=CC(=O)C2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 128.5°C
Color Khaki
Flash Point 141°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Beilstein 07, 724
Fieser 01,714
Merck Index 15, 6480
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alkali hydroxide solutions,freely soluble in hot alcohol,ether,benzene,,chloroform,carbon bisulfide and acetic acid
Formula Weight 158.16
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1, 4-Naphthoquinone, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,4-Naphthoquinone, contains up to 6% moisture
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'