missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Methylbenzyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64420.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15408916

  • £351.92 / 5g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Methylbenzyl)-3-[4-(trifluoromethoxy)phenyl]urea
324.3
MFCD26960700

Specifications

1-(4-Methylbenzyl)-3-[4-(trifluoromethoxy)phenyl]urea
97%
MFCD26960700
Solid
324.3
5g
Videos
SDS