missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxyphenyl)piperazine, 96%, ACROS Organics™

£37.71 - £195.43

Chemical Identifiers

CAS 38212-30-5
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.26
MDL Number MFCD00040736
InChI Key MRDGZSKYFPGAKP-UHFFFAOYSA-N
Synonym 1-4-methoxyphenyl piperazine, n-4-methoxyphenyl piperazine, n-p-methoxyphenyl piperazine, piperazine, 1-4-methoxyphenyl, unii-p385m92gyg, 4-methoxyphenylpiperazine, 1-4-anisyl piperazine, chembl59517, 1-4-methoxyphenyl-piperazine, 1-4-methoxy-phenyl-piperazine
PubChem CID 269722
IUPAC Name 1-(4-methoxyphenyl)piperazine
SMILES COC1=CC=C(C=C1)N1CCNCC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10033240
SDS View Documents
Acros Organics
255830250
25g Glass bottle
£37.71
25g
View alternative products
Save Up to 
»
Estimated Shipment: 11-10-2021
Please to purchase this item. Need a web account? Register with us today!
10718312
SDS View Documents
Acros Organics
255832500
250g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

38212-30-5
192.26
MRDGZSKYFPGAKP-UHFFFAOYSA-N
269722
COC1=CC=C(C=C1)N1CCNCC1
C11H16N2O
MFCD00040736
1-4-methoxyphenyl piperazine, n-4-methoxyphenyl piperazine, n-p-methoxyphenyl piperazine, piperazine, 1-4-methoxyphenyl, unii-p385m92gyg, 4-methoxyphenylpiperazine, 1-4-anisyl piperazine, chembl59517, 1-4-methoxyphenyl-piperazine, 1-4-methoxy-phenyl-piperazine
1-(4-methoxyphenyl)piperazine

Specifications

1-(4-Methoxyphenyl)piperazine
Glass bottle
25g
38212-30-5
100.0
C11H16N2O
1-4-methoxyphenyl piperazine, n-4-methoxyphenyl piperazine, n-p-methoxyphenyl piperazine, piperazine, 1-4-methoxyphenyl, unii-p385m92gyg, 4-methoxyphenylpiperazine, 1-4-anisyl piperazine, chembl59517, 1-4-methoxyphenyl-piperazine, 1-4-methoxy-phenyl-piperazine
COC1=CC=C(C=C1)N1CCNCC1
192.26
192.25
96%
Authentic
Amber to Yellow
96%
95.0
95% min. (GC)
MFCD00040736
MRDGZSKYFPGAKP-UHFFFAOYSA-N
1-(4-methoxyphenyl)piperazine
269722
Crystalline Low Melting Solid
Safety and Handling

Safety and Handling

Product Identifier
  • 1-(4-Methoxyphenyl)piperazine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Product Certifications