missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Methoxyphenyl)-5-methyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2694372
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
187998-64-7
C12H12N2O3
232.239
MFCD02677745
JPFGKGZYCXLEGQ-UHFFFAOYSA-N
2776133
1-(4-methoxyphenyl)-5-methylpyrazole-4-carboxylic acid
CC1=C(C=NN1C2=CC=C(C=C2)OC)C(=O)O
This item is not returnable. View return policy
187998-64-7
C12H12N2O3
232.239
MFCD02677745
JPFGKGZYCXLEGQ-UHFFFAOYSA-N
2776133
1-(4-methoxyphenyl)-5-methylpyrazole-4-carboxylic acid
CC1=C(C=NN1C2=CC=C(C=C2)OC)C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-4-methoxyphenyl-5-methyl-1h-pyrazole-4-carboxylic acid, 1-4-methoxyphenyl-5-methylpyrazole-4-carboxylic acid, 1-4-methoxyphenyl-5-methyl-1h-pyrazole-4-carboxylicacid, 1h-pyrazole-4-carboxylicacid, 1-4-methoxyphenyl-5-methyl, 1h-pyrazole-4-carboxylicacid,1-4-methoxyphenyl-5-methyl
Formula Weight 232.24
Percent Purity 97%
Chemical Name or Material 1-(4-Methoxyphenyl)-5-methyl-1H-pyrazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'