missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Methoxybenzoyl)pyrrolidine, 97%

Brand:  Alfa Aesar™ H59379.03

Code : NEW

Additional Details : CAS Number : 69838-98-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415678

  • £358.49 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Methoxybenzoyl)pyrrolidine
69838-98-8
205.26
MFCD00684253

Specifications

1-(4-Methoxybenzoyl)pyrrolidine
69838-98-8
97%
MFCD00684253
1g
Liquid
205.26
Videos
SDS