missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Isopropylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64357.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15468866

  • £373.12 / 5g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Isopropylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
338.32
MFCD07115606

Specifications

1-(4-Isopropylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
338.32
5g
97%
MFCD07115606
Videos
SDS