missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Iodophenyl)piperazine hydrochloride, 95%

Brand:  Alfa Aesar™ H55299.03

Code : NEW

Additional Details : CAS Number : 624726-35-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15461508

  • £18.76 / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Iodophenyl)piperazine hydrochloride
624726-35-8
324.59
MFCD04119724

Specifications

1-(4-Iodophenyl)piperazine hydrochloride
Solid
324.59
177° to 181°C
Light Sensitive
624726-35-8
95%
MFCD04119724
1g
Videos
SDS