missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Iodophenyl)hydrazine 95%, Maybridge

Brand:  Maybridge RF01374EA

Code : 31

Additional Details : CAS Number : 13116-27-3 Weight : 0.01000kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10680443

  • £242.00 / 10g


Estimated Shipment date
from Supplier 23-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Iodophenyl)hydrazine
95%
IC6H4NHNH2
95%
C6H7IN2
234.04

Specifications

1-(4-Iodophenyl)hydrazine
Amber Glass Bottle
95%
IC6H4NHNH2
10g
13116-27-3
95%
C6H7IN2
234.04
Videos
SDS