missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Iodophenyl)hydrazine 95%, Thermo Scientific™

Catalog Number p-4510349
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
13116-27-3
C6H7IN2
234.04
IQMLCMKMSBMMGR-UHFFFAOYSA-N
4-iodophenyl hydrazine, 1-4-iodophenyl hydrazine, 4-iodo-phenyl-hydrazine, 4-iodo phenyl hydrazine, p-iodophenyl-hydrazine, 4-iodophenyl diazane, pubchem15797, acmc-1brf1, 1-hydrazino-4-iodobenzene, 4-iodophenylhydrazine
704591
(4-iodophenyl)hydrazine
C1=CC(=CC=C1NN)I
This item is not returnable. View return policy
13116-27-3
C6H7IN2
234.04
IQMLCMKMSBMMGR-UHFFFAOYSA-N
4-iodophenyl hydrazine, 1-4-iodophenyl hydrazine, 4-iodo-phenyl-hydrazine, 4-iodo phenyl hydrazine, p-iodophenyl-hydrazine, 4-iodophenyl diazane, pubchem15797, acmc-1brf1, 1-hydrazino-4-iodobenzene, 4-iodophenylhydrazine
704591
(4-iodophenyl)hydrazine
C1=CC(=CC=C1NN)I
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula IC6H4NHNH2
Quantity 1 g
Formula Weight 234.04
Percent Purity 95%
Chemical Name or Material 1-(4-Iodophenyl)hydrazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'