missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1-(4-Fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%
Brand: Alfa Aesar™ H64828.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1-(4-Fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea | |
314.24 | |
MFCD02210841 |
Specifications
1-(4-Fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea | |
Solid | |
314.24 |
1g | |
97% | |
MFCD02210841 |