missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64828.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15489176

  • £351.92 / 5g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
314.24
MFCD02210841

Specifications

1-(4-Fluorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
314.24
5g
97%
MFCD02210841
Videos
SDS