missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Fluorophenyl)-3-(4-isopropylphenyl)urea, 97%

Brand:  Alfa Aesar™ H62746.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419228

  • POA / 1g

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on 01509 555500 for assistance.


Specifications

Chemical Identifiers

1-(4-Fluorophenyl)-3-(4-isopropylphenyl)urea
272.32
MFCD04070347

Specifications

1-(4-Fluorophenyl)-3-(4-isopropylphenyl)urea
Solid
272.32
1g
97%
MFCD04070347
Videos
SDS