missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Fluorophenyl)-3-[3-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H62379.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15407356

  • £66.46 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Fluorophenyl)-3-[3-(trifluoromethyl)phenyl]urea
298.24
MFCD00091657

Specifications

1-(4-Fluorophenyl)-3-[3-(trifluoromethyl)phenyl]urea
Solid
298.24
1g
97%
MFCD00091657
Videos
SDS