missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Fluorophenyl)-3-(3-isopropylphenyl)urea, 97%

Brand:  Alfa Aesar™ H62107.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15428688

  • £367.82 / 5g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Fluorophenyl)-3-(3-isopropylphenyl)urea
272.32
MFCD18175228

Specifications

1-(4-Fluorophenyl)-3-(3-isopropylphenyl)urea
Solid
272.32
5g
97%
MFCD18175228
Videos
SDS