missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Fluorophenyl)-3-[2-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H62084.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15437006

  • £460.04 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Fluorophenyl)-3-[2-(trifluoromethyl)phenyl]urea
298.24
MFCD00091241

Specifications

1-(4-Fluorophenyl)-3-[2-(trifluoromethyl)phenyl]urea
Solid
298.24
5g
97%
MFCD00091241
Videos
SDS