missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Fluorophenyl)-3-(2,3,4-trifluorophenyl)urea, 97%

Brand:  Alfa Aesar™ H62768.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15427826

  • £95.61 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Fluorophenyl)-3-(2,3,4-trifluorophenyl)urea
284.21
MFCD26143338

Specifications

1-(4-Fluorophenyl)-3-(2,3,4-trifluorophenyl)urea
Solid
284.21
1g
97%
MFCD26143338
Videos
SDS