missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Fluorobenzyl)-5-oxo-3-pyrrolidinecarboxylic acid, 97%, Maybridge™

£17.66

Chemical Identifiers

CAS 96449-68-2
Molecular Formula C12H12FNO3
Molecular Weight (g/mol) 237.23
MDL Number MFCD03086215
InChI Key IYAFJFDPMVQTJO-UHFFFAOYSA-N
Synonym 1-4-fluorobenzyl-5-oxopyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxopyrrolidine-3-carboxylic acid, 1-4-fluorobenzyl-5-oxo-3-pyrrolidinecarboxylic acid, 3-pyrrolidinecarboxylicacid, 1-4-fluorophenyl methyl-5-oxo, 1-4-fluorobenzyl-5-oxopyrrolidin-3-carboxylic acid, 1-4-fluorobenzyl-5-oxo-pyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxo-3-pyrrolidinecarboxylic acid, 1-4-fluorophenyl methyl-5-oxo-pyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxidanylidene-pyrrolidine-3-carboxylic acid
PubChem CID 2780702
IUPAC Name 1-[(4-fluorophenyl)methyl]-5-oxopyrrolidine-3-carboxylic acid
SMILES C1C(CN(C1=O)CC2=CC=C(C=C2)F)C(=O)O
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10079391
SDS View Documents
Maybridge
MO00127DA
1g £17.66
1g View alternative products
Save Up to 
»
Estimated Shipment
26-11-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

96449-68-2
237.23
IYAFJFDPMVQTJO-UHFFFAOYSA-N
2780702
C1C(CN(C1=O)CC2=CC=C(C=C2)F)C(=O)O
C12H12FNO3
MFCD03086215
1-4-fluorobenzyl-5-oxopyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxopyrrolidine-3-carboxylic acid, 1-4-fluorobenzyl-5-oxo-3-pyrrolidinecarboxylic acid, 3-pyrrolidinecarboxylicacid, 1-4-fluorophenyl methyl-5-oxo, 1-4-fluorobenzyl-5-oxopyrrolidin-3-carboxylic acid, 1-4-fluorobenzyl-5-oxo-pyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxo-3-pyrrolidinecarboxylic acid, 1-4-fluorophenyl methyl-5-oxo-pyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxidanylidene-pyrrolidine-3-carboxylic acid
1-[(4-fluorophenyl)methyl]-5-oxopyrrolidine-3-carboxylic acid

Specifications

1-(4-Fluorobenzyl)-5-oxo-3-pyrrolidinecarboxylic acid
96449-68-2
97%
MFCD03086215
IYAFJFDPMVQTJO-UHFFFAOYSA-N
1-[(4-fluorophenyl)methyl]-5-oxopyrrolidine-3-carboxylic acid
2780702
97%
1g
97.0
C12H12FNO3
1-4-fluorobenzyl-5-oxopyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxopyrrolidine-3-carboxylic acid, 1-4-fluorobenzyl-5-oxo-3-pyrrolidinecarboxylic acid, 3-pyrrolidinecarboxylicacid, 1-4-fluorophenyl methyl-5-oxo, 1-4-fluorobenzyl-5-oxopyrrolidin-3-carboxylic acid, 1-4-fluorobenzyl-5-oxo-pyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxo-3-pyrrolidinecarboxylic acid, 1-4-fluorophenyl methyl-5-oxo-pyrrolidine-3-carboxylic acid, 1-4-fluorophenyl methyl-5-oxidanylidene-pyrrolidine-3-carboxylic acid
C1C(CN(C1=O)CC2=CC=C(C=C2)F)C(=O)O
237.23
237.23