missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Fluorobenzyl)-1,4-diazepane, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Quantity | 5 g |
Formula Weight | 208.28 |
Percent Purity | 95% |
Chemical Name or Material | 1-(4-Fluorobenzyl)-1, 4-diazepane |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'