missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Fluoro-2-nitrophenyl)azepane, 97%

Brand:  Alfa Aesar™ H63210.03

Code : NEW

Additional Details : Weight : 0.00100kg Transport : UN number : 3259 Chem class : 1b Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15469778

  • £40.60 / 1g


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Specifications

Chemical Identifiers

1-(4-Fluoro-2-nitrophenyl)azepane
238.25
UN3259
1-(4-Fluoro-2-nitrophenyl)hexamethyleneimine

Specifications

1-(4-Fluoro-2-nitrophenyl)azepane
Solid
238.25
1-(4-Fluoro-2-nitrophenyl)hexamethyleneimine
1g
97%
UN3259
Videos
SDS