missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Fluoro-2-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64878.03

UNSPSC : 12352116

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429226

  • £56.92 / 1g


Estimated Shipment date
from Supplier 07-07-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

1-(4-Fluoro-2-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
328.26
MFCD26960712

Specifications

1-(4-Fluoro-2-methylphenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
328.26
1g
97%
MFCD26960712
Documentation

Technical Documents