missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-dioxaspiro[4.5]decan-8-one, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Formula Weight | 156.18 |
Chemical Name or Material | 1, 4-Dioxaspiro[4.5]decan-8-one |
Safety and Handling
Signal Word
- Warning
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H335-May cause respiratory irritation.
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Cyclohexane-1,4-dione, monoethylene ketal
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'