missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxaspiro[4.5]decan-8-ol, 98%, Thermo Scientific Chemicals

Catalog Number p-3736369
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
22428-87-1
C8H14O3
158.20
MFCD00067003
HKQTYQDNCKMNHZ-UHFFFAOYSA-N
1,4-dioxaspiro 4.5 decan-8-ol, 1,4-dioxa-spiro 4.5 decan-8-ol, 4-hydroxycyclohexanone ethylene acetal, 4-hydroxycyclohexanone monoethylene ketal, 8-hydroxy-1,4-dioxaspiro 4.5 decane, pubchem20110, acmc-1chv5, 4,4-ethylenedioxycyclohexanol
567352
1,4-dioxaspiro[4.5]decan-8-ol
OC1CCC2(CC1)OCCO2
This item is not returnable. View return policy
22428-87-1
C8H14O3
158.20
MFCD00067003
HKQTYQDNCKMNHZ-UHFFFAOYSA-N
1,4-dioxaspiro 4.5 decan-8-ol, 1,4-dioxa-spiro 4.5 decan-8-ol, 4-hydroxycyclohexanone ethylene acetal, 4-hydroxycyclohexanone monoethylene ketal, 8-hydroxy-1,4-dioxaspiro 4.5 decane, pubchem20110, acmc-1chv5, 4,4-ethylenedioxycyclohexanol
567352
1,4-dioxaspiro[4.5]decan-8-ol
OC1CCC2(CC1)OCCO2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4850 to 1.487
Quantity 1 g
Formula Weight 158.2
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 1, 4-Dioxaspiro[4.5]decan-8-ol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'