missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dioxane, Reag. Ph. Eur. reag. ISO, Honeywell
Click to view available options
Quantity:
1 L
2.5 L
210 kg
Specifications
Density | 1.034g/mL at 25°C |
Quantity | 1 L |
Explosion Limit | 0.22 |
Flash Point | 12°C (53.6°F) |
Ignition Point | 356°F |
Vapor Pressure | 40mmHg (25°C), 27mmHg (20°C) |
UN Number | 1165 |
Beilstein | 102551 |
Formula Weight | 88.11g/mol |
Percent Purity | ≥99.5% (GC) |
Show More |
Safety and Handling
Product Identifier
- 1,4-dioxane
- Danger
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- EUH019-May form explosive peroxides.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : 1,4-dioxane
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'