missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, HPLC Grade, 99% min, Thermo Scientific Chemicals

Catalog Number p-4408249
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
123-91-1
C4H8O2
88.106
MFCD00006571
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1
123-91-1
C4H8O2
88.106
MFCD00006571
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1

1,4-Dioxane HPLC Grade is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 11.8°C
Color Colorless
Appearance Colorless liquid
Density 1.034
Boiling Point 100°C to 102°C
Quantity 1 L
Flash Point 12°C (54°F)
Ignition Point 375°C (707°F)
Odor Ether like
Physical Form Liquid
Refractive Index 1.422
Assay Percent Range 99% min.
Vapor Pressure 41 hPa (31mm Hg) at 20°C
UN Number 1165
Beilstein 102551
Solubility Information Miscible with water,most organic solvents,aromatic hydrocarbons and oils.
Formula Weight 88.11
Grade HPLC
Concentration or Composition (by Analyte or Components) 1, 4-Dioxane
Chemical Name or Material 1, 4-Dioxane, Filtered through 0.2μ filters.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'