missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, anhydrous, 99.8%, stab. with 1-3ppm BHT, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog Number 11330478
Click to view available options
Quantity:
100 mL
1 L
4 x 1 L
123-91-1
C4H8O2
88.106
MFCD00006571
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1
123-91-1
C4H8O2
88.106
MFCD00006571
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1

1,4-Dioxane is used as a stabilizer for 1,1,1-trichloroethane and chlorinated solvents. It acts as a solvent for cellulose acetate, ethyl cellulose, benzyl cellulose, resins, oils, waxes, dyes, other organic and inorganic compounds. It acts as a substitute for tetrahydrofuran due to its higher boiling point and lower toxicity. Further, it is used as an internal standard for proton NMR spectroscopy in deuterium oxide. It plays an important role in liquid scintillation counting in biological procedures, and as a wetting and dispersing agent in textile processing. It is involved in the preparation of dimethylmagnesium by reacting with methylmagnesium bromide, and in the preparation of histological sections for microscopic examination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 11.8°C
Color Colorless
Appearance Colorless liquid
Density 1.034
Boiling Point 100°C to 102°C
Quantity 1 L
Flash Point 12°C (54°F)
Ignition Point 375°C (707°F)
Odor Ether like
Physical Form Liquid
Refractive Index 1.422
Assay Percent Range 99.8%
Vapor Pressure 41 hPa (31mm Hg) at 20°C
UN Number 1165
Beilstein 102551
Solubility Information Miscible with water,most organic solvents,aromatic hydrocarbons and oils.
Formula Weight 88.11
Concentration or Composition (by Analyte or Components) 1, 4-Dioxane, anhydrous
Chemical Name or Material 1, 4-Dioxane, anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'