missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, anhydrous, 99.8%, Alfa Aesar™

Brand:  Alfa Aesar 043821.M1

Code : NEW

Additional Details : CAS Number : 123-91-1 Weight : 0.01000kg Transport : UN number : 1165 Chem class : 3 Pack group : II

Quantity 4 x 1L

 View more versions of this product

Product Code. 11320478

  • £217.00 / 4 L

Estimated Shipment: 08-10-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

123-91-1
88.106
RYHBNJHYFVUHQT-UHFFFAOYSA-N
31275
1,4-dioxane
C4H8O2
MFCD00006571
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
CHEBI:47032
C1COCCO1

Specifications

1, 4-Dioxane, anhydrous
123-91-1
99.8%
MFCD00006571
102551
Miscible with water,most organic solvents,aromatic hydrocarbons and oils.
C1COCCO1
88.106
CHEBI:47032
Liquid
375°C (707°F)
1.422
12°C (54°F)
Colorless
4 x 1L
1, 4-Dioxane, anhydrous
Colorless liquid
C4H8O2
1165
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
RYHBNJHYFVUHQT-UHFFFAOYSA-N
1,4-dioxane
31275
88.11
1.034
Ether like
41 hPa (31mm Hg) at 20°C
100°C to 102°C
11.8°C
Hygroscopic
Videos
SDS Documents
Certificates