missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, Reagent Grade, ≥99%, Honeywell™

Catalog Number p-7120018
Click to view available options
Quantity:
500 mL
2.5 L
10 L
20 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
123-91-1
C4H8O2
88.106
MFCD00006571
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1
This item is not returnable. View return policy
123-91-1
C4H8O2
88.106
MFCD00006571
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
pH 6 to 8 (at 500.00g/L 20°C)
Melting Point 12°C
Color Colorless
UN Number UN1165
Beilstein 102551
Solubility Information Fully miscible in water
Formula Weight 88.11g/mol
Percent Purity ≥99%
Density 1.034g/mL at 25°C
Boiling Point 100°C to 102°C (1,013 hPa)
Quantity 500 mL
Explosion Limit 0.22
Flash Point 12°C (53.6°F)
Ignition Point 356°F
Odor Aromatic
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 27mmHg (20°C), 40mmHg (25°C)
Chemical Name or Material 1,4-Dioxane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'