missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dioxane, 99%, for biochemistry, stabilized, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4440169
52.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
123-91-1
C4H8O2
88.106
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1
123-91-1
C4H8O2
88.106
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 12.0°C
Absorbance 0.05 max. at 295nm, 0.1 max. at 275nm, 0.5 max. at 250nm
Density 1.0340g/mL
Boiling Point 101.0°C
Quantity 1 L
Flash Point 12°C
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4205 to 1.4220
Assay Percent Range 99%
Specific Gravity 1.034
Viscosity 1.32 mPa.s (20°C)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents
Formula Weight 88.11
Percent Purity 99%
Grade Biochemical
Chemical Name or Material 1, 4-Dioxane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'