missing translation for 'onlineSavingsMsg'
Learn More

1,4-dimethylpiperazine-2-carboxylic acid dihydrochloride, Thermo Scientific™

Catalog Number p-4391328
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
1174064-55-1
C7H16Cl2N2O2
231.117
UPJQXMOMHHDOSZ-UHFFFAOYSA-N
1,4-dimethylpiperazine-2-carboxylic acid dihydrochloride, 1,4-dimethylpiperazine-2-carboxylic acid,dihydrochloride, 1,4-dimethylpiperazine-2-carboxylic acid, chloride, chloride, 1,4-dimethylpiperazine-2-carboxylic acid-hydrogen chloride 1/2
45594293
1,4-dimethylpiperazine-2-carboxylic acid;dihydrochloride
CN1CCN(C(C1)C(=O)O)C.Cl.Cl
This item is not returnable. View return policy
1174064-55-1
C7H16Cl2N2O2
231.117
UPJQXMOMHHDOSZ-UHFFFAOYSA-N
1,4-dimethylpiperazine-2-carboxylic acid dihydrochloride, 1,4-dimethylpiperazine-2-carboxylic acid,dihydrochloride, 1,4-dimethylpiperazine-2-carboxylic acid, chloride, chloride, 1,4-dimethylpiperazine-2-carboxylic acid-hydrogen chloride 1/2
45594293
1,4-dimethylpiperazine-2-carboxylic acid;dihydrochloride
CN1CCN(C(C1)C(=O)O)C.Cl.Cl
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 231.12
Percent Purity 95%
Chemical Name or Material 1, 4-dimethylpiperazine-2-carboxylic acid dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'