missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diisopropylbenzene, 99%, Thermo Scientific Chemicals

Catalog Number p-113216
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
100-18-5
C12H18
162.28
MFCD00008892
SPPWGCYEYAMHDT-UHFFFAOYSA-N
1,4-diisopropylbenzene, p-diisopropylbenzene, benzene, p-diisopropyl, benzene, 1,4-bis 1-methylethyl, para-diisopropylbenzene, p-diisopropylbenzol, 1,4-bis 1-methylethyl benzene, unii-5m62031nzp, 1,4-di propan-2-yl benzene, dsstox_cid_6652
7486
CC(C)C1=CC=C(C=C1)C(C)C
This item is not returnable. View return policy
100-18-5
C12H18
162.28
MFCD00008892
SPPWGCYEYAMHDT-UHFFFAOYSA-N
1,4-diisopropylbenzene, p-diisopropylbenzene, benzene, p-diisopropyl, benzene, 1,4-bis 1-methylethyl, para-diisopropylbenzene, p-diisopropylbenzol, 1,4-bis 1-methylethyl benzene, unii-5m62031nzp, 1,4-di propan-2-yl benzene, dsstox_cid_6652
7486
CC(C)C1=CC=C(C=C1)C(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -17.0°C
Density 0.8600g/mL
Boiling Point 203.0°C
Flash Point 76°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4[CH(CH3)2]2
Refractive Index 1.4890 to 1.4910
Quantity 5 g
Beilstein 05,II,339
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble. Other solubilities: %iscible with alcohol, ether, benzene, acetone and, ccl4
IUPAC Name 1,4-bis(propan-2-yl)benzene
Formula Weight 162.27
Percent Purity 99%
Chemical Name or Material 1,4-Diisopropylbenzene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Diisopropylbenzene
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'