missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diisocyanatobutane, 97%, Thermo Scientific Chemicals

Catalog Number p-3757721
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
4538-37-8
C6H8N2O2
140.14
MFCD00075559
OVBFMUAFNIIQAL-UHFFFAOYSA-N
350453
1,4-diisocyanatobutane
C(CCN=C=O)CN=C=O
This item is not returnable. View return policy
4538-37-8
C6H8N2O2
140.14
MFCD00075559
OVBFMUAFNIIQAL-UHFFFAOYSA-N
350453
1,4-diisocyanatobutane
C(CCN=C=O)CN=C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.1000g/mL
Boiling Point 102.0°C to 104.0°C (14.0 mmHg)
Flash Point 106°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula OCN(CH2)4NCO
Refractive Index 1.4510 to 1.4530
Quantity 1 g
UN Number 2206
Specific Gravity 1.1
Synonym 1,4-butanediisocyanate, tetramethylene diisocyanate, 1,4-disocyanatobutane, acmc-1afua, 1,4-diisocyanato-butane, butane-1,4-diisocyanate, #, butane,1,4-diisocyanato, 1,4-butanediyl diisocyanate, butane, 1,4-diisocyanato
Formula Weight 140.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 4-Diisocyanatobutane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'