missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dihydroxy-2-naphthoic acid, 98%, Thermo Scientific™

Catalog Number p-7058087
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
31519-22-9
C11H8O4
204.18
MFCD00010370
VOJUXHHACRXLTD-UHFFFAOYSA-N
1,4-dihydroxy-2-naphthoic acid, 1,4-dihydroxy-2-naphthalenecarboxylic acid, 1,4-dihydroxy-2-naphthoate, 2-naphthalenecarboxylic acid, 1,4-dihydroxy, unii-1u77mr5q7o, 1,4-dihydroxy-2-naphtoic acid, 1,4-dohydroxy-2-naphthoic acid, 1,4-dihydroxy-2-naphthalic acid, cbmicro_022187, acmc-209hn7
671
CHEBI:18094
1,4-dihydroxynaphthalene-2-carboxylic acid
OC(=O)C1=CC(O)=C2C=CC=CC2=C1O
This item is not returnable. View return policy
31519-22-9
C11H8O4
204.18
MFCD00010370
VOJUXHHACRXLTD-UHFFFAOYSA-N
1,4-dihydroxy-2-naphthoic acid, 1,4-dihydroxy-2-naphthalenecarboxylic acid, 1,4-dihydroxy-2-naphthoate, 2-naphthalenecarboxylic acid, 1,4-dihydroxy, unii-1u77mr5q7o, 1,4-dihydroxy-2-naphtoic acid, 1,4-dohydroxy-2-naphthoic acid, 1,4-dihydroxy-2-naphthalic acid, cbmicro_022187, acmc-209hn7
671
CHEBI:18094
1,4-dihydroxynaphthalene-2-carboxylic acid
OC(=O)C1=CC(O)=C2C=CC=CC2=C1O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,4-Dihydroxy-2-naphthoic acid from Propionibacterium freudenreichii is known to promote the proliferation of Bifidobacterium. It has potential therapeutic application for psoriasis treatment.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 220°C (decomposition)
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 204.18
Percent Purity 98%
Chemical Name or Material 1,4-Dihydroxy-2-naphthoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'