missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diethylbenzene, 98%, Thermo Scientific Chemicals

Catalog Number p-107932
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
105-05-5
C10H14
134.22
MFCD00009264
DSNHSQKRULAAEI-UHFFFAOYSA-N
p-diethylbenzene, benzene, 1,4-diethyl, p-ethylethylbenzene, benzene, p-diethyl, para-diethylbenzene, unii-0psm16x42d, acmc-1bu3z, dsstox_cid_6711, dsstox_rid_78192, dsstox_gsid_26711
7734
CHEBI:34062
1,4-diethylbenzene
CCC1=CC=C(CC)C=C1
This item is not returnable. View return policy
105-05-5
C10H14
134.22
MFCD00009264
DSNHSQKRULAAEI-UHFFFAOYSA-N
p-diethylbenzene, benzene, 1,4-diethyl, p-ethylethylbenzene, benzene, p-diethyl, para-diethylbenzene, unii-0psm16x42d, acmc-1bu3z, dsstox_cid_6711, dsstox_rid_78192, dsstox_gsid_26711
7734
CHEBI:34062
1,4-diethylbenzene
CCC1=CC=C(CC)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -43°C
Color Colorless
Density 0.8600g/mL
Boiling Point 184°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(C2H5)2
Refractive Index 1.4935 to 1.4955
Quantity 5 mL
Beilstein 05, 426
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in benzene and carbon tetrachloride,miscible with ethanol,ether and acetone
Formula Weight 134.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 4-Diethylbenzene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Diethylbenzene
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'