missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals
22.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Decomposition Information | >173°C |
Melting Point | 52°C to 56°C |
Color | White |
Density | 1.2400g/mL |
Boiling Point | 174°C |
Flash Point | 67°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Show More |
Safety and Handling
Product Identifier
- 1,4-Dichlorobenzene
- Warning
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H410-Very toxic to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'