missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichlorobenzene, 99+%, Thermo Scientific Chemicals

Catalog Number p-103965
22.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
106-46-7
C6H4Cl2
147
MFCD00000604
OCJBOOLMMGQPQU-UHFFFAOYSA-N
p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola
4685
CHEBI:28618
1,4-dichlorobenzene
C1=CC(=CC=C1Cl)Cl
This item is not returnable. View return policy
106-46-7
C6H4Cl2
147
MFCD00000604
OCJBOOLMMGQPQU-UHFFFAOYSA-N
p-dichlorobenzene, paradichlorobenzene, para-dichlorobenzene, paracide, paramoth, dichlorobenzene, paranuggets, santochlor, paradow, evola
4685
CHEBI:28618
1,4-dichlorobenzene
C1=CC(=CC=C1Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Decomposition Information >173°C
Melting Point 52°C to 56°C
Color White
Density 1.2400g/mL
Boiling Point 174°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula C6H4Cl2
Quantity 1 kg
Beilstein 05, 203
Merck Index 15, 3066
Specific Gravity 1.24
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,benzene,ether,chloroform,and carbon disulfide
Formula Weight 147
Vapor Pressure 1.7mbar at 20°C
Percent Purity 99+%
Physical Form Crystals
Chemical Name or Material 1, 4-Dichlorobenzene, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,4-Dichlorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'