missing translation for 'onlineSavingsMsg'
Learn More

1,4-Dichloro-2-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog Number p-7081763
Click to view available options
Quantity:
100 g
250 g
500 g
2500 g
This item is not returnable. View return policy
89-61-2
C6H3Cl2NO2
192.00
MFCD00007074
RZKKOBGFCAHLCZ-UHFFFAOYSA-N
2,5-dichloronitrobenzene, nitro-p-dichlorobenzene, benzene, 1,4-dichloro-2-nitro, 1-nitro-2,5-dichlorobenzene, 2,5-dichlornitrobenzen, 2,5-dichloro-1-nitrobenzene, benzene, 2,5-dichloronitro, 2,5-dichloronitrobenzol, unii-iy18g50ff4, ccris 3096
6977
1,4-dichloro-2-nitrobenzene
[O-][N+](=O)C1=CC(Cl)=CC=C1Cl
This item is not returnable. View return policy
89-61-2
C6H3Cl2NO2
192.00
MFCD00007074
RZKKOBGFCAHLCZ-UHFFFAOYSA-N
2,5-dichloronitrobenzene, nitro-p-dichlorobenzene, benzene, 1,4-dichloro-2-nitro, 1-nitro-2,5-dichlorobenzene, 2,5-dichlornitrobenzen, 2,5-dichloro-1-nitrobenzene, benzene, 2,5-dichloronitro, 2,5-dichloronitrobenzol, unii-iy18g50ff4, ccris 3096
6977
1,4-dichloro-2-nitrobenzene
[O-][N+](=O)C1=CC(Cl)=CC=C1Cl

1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.

Solubility
Soluble in water, ethanol, ether, benzene, carbon disulfide. Slightly soluble in carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 53°C to 57°C
Boiling Point 266°C to 269°C
Flash Point 125°C (257°F)
Quantity 100 g
UN Number UN3077
Beilstein 778109
Formula Weight 192
Percent Purity 98%
Chemical Name or Material 1,4-Dichloro-2-nitrobenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'