missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diazepane, 97%, Maybridge

Brand:  Maybridge SB01866EB

Code : 31

Additional Details : CAS Number : 505-66-8 Weight : 0.07500kg Transport : UN number : 3259 Chem class : 1b Pack group : III

Quantity 25g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10133124

  • £22.00 / 25g


Estimated Shipment date
from Supplier 29-03-2018
Add to basket

Specifications

Chemical Identifiers

1, 4-Diazepane
95.0
95%
100.16
505-66-8
95%
C5H12N2
1, 4-Diazacycloheptane

Specifications

1, 4-Diazepane
95.0
95%
25g
100.16
505-66-8
Amber Glass Bottle
95%
C5H12N2
1, 4-Diazacycloheptane
Videos
SDS