missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,4-Diacryloylpiperazine, Electrophoresis Reagent, 97%

Brand:  Alfa Aesar™ J66421.14

Code : NEW

Additional Details : Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15430587

  • £385.84 / 25g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

1,4-Diacryloylpiperazine
Electrophoresis Reagent
MFCD00077661
1,4-Bis(acryloyl)piperazine; Diacroyl piperazine
6342-17-2
194.23
745431

Specifications

1,4-Diacryloylpiperazine
Electrophoresis Reagent
Powder
194.23
745431
White to off-white
25g
6342-17-2
97%
MFCD00077661
1,4-Bis(acryloyl)piperazine; Diacroyl piperazine
91° to 95°C
Videos
SDS