Alfa Aesar™ 1,4-Diacryloylpiperazine, Electrophoresis Reagent, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: J66421.06

5GR 1,4-Diacryloylpiperazine, Electrophoresis Reagent, 97% 5g

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.00500kg


Quantity 5g

Product Code. 15420587

Quantity Price
1 £ 101.12 / 5g
Estimated Shipment date
from Supplier 01-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1,4-Diacryloylpiperazine
CAS 6342-17-2
Name Note Electrophoresis Reagent
Formula Weight 194.23
MDL Number MFCD00077661
Beilstein 745431
Synonym 1,4-Bis(acryloyl)piperazine; Diacroyl piperazine

Specification

Formula Weight 194.23
Melting Point 91° to 95°C
Chemical Name or Material 1,4-Diacryloylpiperazine
Name Note Electrophoresis Reagent
Physical Form Powder
Assay 97%
MDL Number MFCD00077661
CAS 6342-17-2
Beilstein 745431
Synonym 1,4-Bis(acryloyl)piperazine; Diacroyl piperazine
Color White to off-white
Quantity 5g