missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cineole, 85%, Thermo Scientific Chemicals

Catalog Number p-7077598
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
470-67-7
C10H18O
154.25
RFFOTVCVTJUTAD-UHFFFAOYSA-N
1,4-cineole, isocineole, 1,4-cineol, 1,4-epoxy-p-menthane, isocineple, 1,4-cineole natural, p-menthane, 1,4-epoxy, unii-b55jtu839b, fema no. 3658
10106
CHEBI:80788
1-methyl-4-propan-2-yl-7-oxabicyclo[2.2.1]heptane
CC(C)C12CCC(O1)(CC2)C
This item is not returnable. View return policy
470-67-7
C10H18O
154.25
RFFOTVCVTJUTAD-UHFFFAOYSA-N
1,4-cineole, isocineole, 1,4-cineol, 1,4-epoxy-p-menthane, isocineple, 1,4-cineole natural, p-menthane, 1,4-epoxy, unii-b55jtu839b, fema no. 3658
10106
CHEBI:80788
1-methyl-4-propan-2-yl-7-oxabicyclo[2.2.1]heptane
CC(C)C12CCC(O1)(CC2)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -46.0°C
Density 0.8870g/mL
Boiling Point 65°C (21.0 hPa)
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 80% min. (GC)
Quantity 25 g
Specific Gravity 0.887
Formula Weight 154.25
Percent Purity 85%
Chemical Name or Material 1,4-Cineole
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Cineole
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'