missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Cineole, 85%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Melting Point | -46.0°C |
Density | 0.8870g/mL |
Boiling Point | 65°C (21.0 hPa) |
Flash Point | 47°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 80% min. (GC) |
Quantity | 25 g |
Specific Gravity | 0.887 |
Formula Weight | 154.25 |
Percent Purity | 85% |
Mostrar más |
Seguridad y manipulación
Nombre del producto
- 1,4-Cineole
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'