missing translation for 'onlineSavingsMsg'
Learn More

1-[(4-chlorophenyl)sulfonyl]acetone, Thermo Scientific™

Catalog Number p-4391262
Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
5000-48-6
C9H9ClO3S
232.68
MFCD00018663
BRDBHPZILGTBFY-UHFFFAOYSA-N
4-chlorophenylsulfonylacetone, 1-4-chlorophenyl sulfonyl acetone, 1-4-chlorophenyl sulfonyl propan-2-one, 4-chlorobenzenesulphonylacetone, p-chlorophenylsulfonylacetone, 1-4-chlorobenzenesulfonyl propan-2-one, 4-chlorophenylsulfonyl acetone, 1-4-chlorophenyl sulfonylpropan-2-one, maybridge1_004685
521269
1-(4-chlorophenyl)sulfonylpropan-2-one
CC(=O)CS(=O)(=O)C1=CC=C(Cl)C=C1
This item is not returnable. View return policy
5000-48-6
C9H9ClO3S
232.68
MFCD00018663
BRDBHPZILGTBFY-UHFFFAOYSA-N
4-chlorophenylsulfonylacetone, 1-4-chlorophenyl sulfonyl acetone, 1-4-chlorophenyl sulfonyl propan-2-one, 4-chlorobenzenesulphonylacetone, p-chlorophenylsulfonylacetone, 1-4-chlorobenzenesulfonyl propan-2-one, 4-chlorophenylsulfonyl acetone, 1-4-chlorophenyl sulfonylpropan-2-one, maybridge1_004685
521269
1-(4-chlorophenyl)sulfonylpropan-2-one
CC(=O)CS(=O)(=O)C1=CC=C(Cl)C=C1
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 232.68
Percent Purity 97%
Chemical Name or Material 1-[(4-chlorophenyl)sulfonyl]acetone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'