missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-chlorophenyl)ethanol, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Density | 1.1700g/mL |
Boiling Point | 136°C (2.7 kPa) |
Flash Point | >110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Quantity | 25 g |
Specific Gravity | 1.17 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'