missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid, 97%, Maybridge™

Brand:  Maybridge GK01550EA

Code : 31

Additional Details : CAS Number : 98534-80-6 Weight : 0.06000kg

Packaging Amber Glass Bottle
Quantity 10g

 View more versions of this product

Product Code. 10005962

  • £56.26 / 10g

Estimated Shipment
01-06-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

98534-80-6
290.626
HSZJMDBOSZVDOK-UHFFFAOYSA-N
688687
C1=CC(=CC=C1N2C(=C(C=N2)C(=O)O)C(F)(F)F)Cl
C11H6ClF3N2O2
MFCD00068142
1-4-chlorophenyl-5-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 2-4-chlorophenyl-3-trifluoromethyl pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-trifluoromethyl pyrazole-4-carboxylic acid, 2-4-chlorophenyl-3-trifluoromethyl-2h-pyrazole-4-carboxylic acid, maybridge1_004008, 2-4-chlorophenyl-3-trifluoromethyl pyrazole-4, 1-4-chlorophenyl-5-trifluoromethyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-trifluoromethyl
1-(4-chlorophenyl)-5-(trifluoromethyl)pyrazole-4-carboxylic acid

Specifications

1-(4-Chlorophenyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
10g
96.5
97%
MFCD00068142
HSZJMDBOSZVDOK-UHFFFAOYSA-N
1-(4-chlorophenyl)-5-(trifluoromethyl)pyrazole-4-carboxylic acid
688687
97%
Amber Glass Bottle
98534-80-6
100.0
C11H6ClF3N2O2
1-4-chlorophenyl-5-trifluoromethyl-1h-pyrazole-4-carboxylic acid, 2-4-chlorophenyl-3-trifluoromethyl pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-trifluoromethyl pyrazole-4-carboxylic acid, 2-4-chlorophenyl-3-trifluoromethyl-2h-pyrazole-4-carboxylic acid, maybridge1_004008, 2-4-chlorophenyl-3-trifluoromethyl pyrazole-4, 1-4-chlorophenyl-5-trifluoromethyl-4-pyrazolecarboxylic acid, 1h-pyrazole-4-carboxylic acid, 1-4-chlorophenyl-5-trifluoromethyl
C1=CC(=CC=C1N2C(=C(C=N2)C(=O)O)C(F)(F)F)Cl
290.626
290.63
Videos
SDS Documents